missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG H and L Chain Goat anti-Human, FITC, Polyclonal, MP Biomedicals
Goat Polyclonal Antibody
Brand: MP Biomedicals 08672161
Additional Details : Weight : 0.01000kg
Description
IgG H and L Chain Polyclonal antibody specifically detects IgG H and L Chain in Human samples. It is validated for Immunofluorescence.Specifications
IgG H and L Chain | |
Polyclonal | |
Human | |
2 mg | |
Secondary | |
Store at 0° to 5°C |
Immunofluorescence | |
FITC | |
Affinity Purified | |
H and L Chain | |
Human | |
Lyophilized |