Learn More
Invitrogen™ IGF-2 Mouse ELISA Kit
Sandwich ELISA Kit
626.00 EUR - 2635.00 EUR
Specifications
Accession Number | P09535 |
---|---|
Assay Sensitivity | 1500 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15445283
|
Invitrogen™
EMIGF2 |
96 Tests |
626.00 EUR
1 set |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
17145739
|
Invitrogen™
EMIGF2X5 |
5 x 96 Tests |
2635.00 EUR
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The Mouse Insulin-like Growth Factor 2 (Ms IGF-2) ELISA quantitates Ms IGF-2 in mouse cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IGF-2. Principle of the method The Mouse IGF-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The insulin gene family, comprised of insulin, relaxin and insulin-like growth factors I and II (IGF-I and IGF-II), represents a group of structurally related polypeptides whose biological functions have diverged. The IGFs, or somato-medins, constitute a class of polypeptides that have a key role in pre-adolescent mammalian growth. IGF-I and -II are critical regulators of cell proliferation and differentiation. Most of the growth promoting properties of both ligands are mediated by the IGF-I receptor (IGF-IR). IGF- I and -II, respectively known as somatomedin C and somatomedin A, are single chain polypeptides which share an amino acid sequence homology of about 47% with insulin. IGF-I expression is regulated by growth hormone and mediates postnatal growth, while IGF-II is induced by placental lactogen during prenatal development. IGF-II is a fetal growth factor, influenced by placental lactogen and abundantly expressed by placental trophoblasts. IGF-II and IGF-binding protein 1 (IGFBP1) gene variants are associated with overfeeding-induced metabolic changes. The human IGF-II gene maps to chromosome 11p15.5, encoding a 180 amino acid protein which is the precursor to IGF-II.Specifications
P09535 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Supernatant | |
Mouse | |
Colorimetric Microplate Reader | |
AL033362,Igf-2,Igf-II,M6pr,Mpr,Peg2 | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
1500 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
The antibody pair provided in this kit recognizes mouse IGF-II (for Cell Culture). | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
16002 | |
<12% | |
HRP | |
Development | |
AL033362, Igf-2, Igf-II, M6pr, Mpr, Peg2 | |
1 hr. 20 min. |