Learn More
IgA (α-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody, Epredia™
IgA (α-Heavy Chain) Ab-2, Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
158.10 EUR - 243.00 EUR
Specifications
Antigen | IgA (alpha-Heavy Chain) Ab-2 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12672887
|
Epredia
RB-1433-A1 |
Concentrated | 500 μL |
243.00 EUR
500µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12662887
|
Epredia
RB-1433-A0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12652887
|
Epredia
RB-1433-A |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12602897
|
Epredia
RB-1433-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: IgA isolated from a pool of normal human sera
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic Ab-2 recognizes a protein of 50kDa, identified as alpha heavy chain of human immunoglobulins. It reacts with both IgA1 and IgA2 isotypes. Ab-2 does not cross-react with γ (IgG), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
IgA (alpha-Heavy Chain) Ab-2 | |
Polyclonal | |
Rabbit | |
Immunoglobulins | |
IgA isolated from a pool of normal human sera | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG |