Learn More
IgA (α-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody, Epredia™
IgA (α-Heavy Chain) Ab-2, Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Brand: Epredia RB-1433-A1
Additional Details : Weight : 0.00190kg
Description
The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: IgA isolated from a pool of normal human sera
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic Ab-2 recognizes a protein of 50kDa, identified as alpha heavy chain of human immunoglobulins. It reacts with both IgA1 and IgA2 isotypes. Ab-2 does not cross-react with γ (IgG), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
IgA (alpha-Heavy Chain) Ab-2 | |
Polyclonal | |
Rabbit | |
500 μL | |
Immunoglobulins | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IgA isolated from a pool of normal human sera | |
IVD | |
Primary | |
Concentrated |