missing translation for 'onlineSavingsMsg'
Learn More

IgA (α-Heavy Chain) Ab-2 Rabbit Polyclonal Antibody, Epredia™

IgA (α-Heavy Chain) Ab-2, Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia RB-1433-A0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12662887

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: IgA isolated from a pool of normal human sera

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic Ab-2 recognizes a protein of 50kDa, identified as alpha heavy chain of human immunoglobulins. It reacts with both IgA1 and IgA2 isotypes. Ab-2 does not cross-react with γ (IgG), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. Ab-2 is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

IgA (alpha-Heavy Chain) Ab-2
Polyclonal
Rabbit
100 μL
Immunoglobulins
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
IgA isolated from a pool of normal human sera
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers