Learn More
Brady™ i3300 Industrial Label Printer - EU with Brady Workstation LAB Suite
The BradyPrinter i3300 Industrial Label Printer can print a huge range of sign and labels in monocolour. Print labels to identify wires, products, laboratory samples and facilities without any hassle.
Brand:  Brady™ 241131
1372.75 EUR valid until 2024-12-31
Use promo code "21918" to get your promotional price.
Additional Details : Weight : 7.51000kg
 View more versions of this product
Product Code. 16306935
  • 1445.00 EUR / Each
Estimated Shipment: 26-06-2024
Description
Get your ideal label printing scenario with the BradyPrinter i3300 Industrial Label printer – labels and ribbons that snap into place, no calibration and no wasted blank labels.
  • Mid-volume industrial label printer ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels
  • Print smart technology allows the printer to print on the first label – even sleeves and small labels
  • Changeovers in a flash – foolproof drop-in label rolls and ribbons snap into place in 20 seconds
  • Talk about options, this printer prints on more than 825 parts and 70 material options for facility-wide identification
  • Ships with Brady Workstation PWID Software Suites to your solution and make label creation even easier
  • Setup is stress free – Contact our Brady Technical Support Team and we will schedule a free phone setup appointment
Specifications
Specifications
Color LCD Touchscreen
305 x 231 mm
With Wifi
101.60 mm/sec.
100/240 V, 50/60 Hz
Thermal Transfer
Monochrome
This i3300 printer comes in a cardboard box. It ships with a power cord, a Brady Workstation LAB Suite, a USB cable, a product CD holding the manual, a cleaning kit and a printed quick start guide
300 dpi