missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Campylobacter jejuni Mouse anti-Bacteria, Clone: HM035, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13496.1mg
Additional Details : Weight : 0.02000kg
Description
Mouse monoclonal antibody raised against Campylobacter jejuni.
- High specificity
- High reactivity
Specifications
Campylobacter jejuni | |
Monoclonal | |
Unconjugated | |
ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Bacteria | |
Liquid |
ELISA, Immunofluorescence, Lateral Flow | |
HM035 | |
Mouse monoclonal antibody raised against Campylobacter jejuni. | |
In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
Protein A purification | |
RUO | |
This antibody reacts with whole cells. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
For Research Use Only (RUO)