missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-928 Medium Duty Gloves, Glass-Free INTERCEPT™ Liner

Medium duty gloves with glass-free INTERCEPT liner provides dual protection from cuts and mechanical fluids.

Brand:  Ansell 11928100

Additional Details : Weight : 6.62400kg

 View more versions of this product

Product Code. 16029901

  • 190.00 EUR / 12 pairs

Estimated Shipment: 15-02-2023
to see stock.
Add to basket

Description

Description

  • Silicone and DMF free
  • Grip performance for safe handling in dry to light oily applications
  • Double dipped nitrile coating protects against oil and industrial fluid exposure while providing high durability and extended use life
  • INTERCEPT Aramid Liner for increased cut performance
  • Contact Heat Protection Level 1
Specifications

Specifications

ANSI 3
Nitrile
ANSI A4
Dip-Coated Gloves
EN 388:4X32D
Knit
11928100
9.3 in.
CE 0086, EN420:2003 + A1:2009; REACH Compliant; Category II; EN407 X1XXXX
Black
Yes
10
Automotive, metal fabrication, machinery and equipment
Ansell Intercept™ Technology, Aramid
HyFlex™ 11-928
23.6 cm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers