Learn More
Ansell Edmont™ HyFlex™ Lightweight Polyurethane Gloves
Water-based polyurethane increases dexterity and comfort.
Brand: Ansell Edmont™ 11-402_11
Additional Details : Weight : 0.01000kg
Description
- Seamless construction
- Abrasion resistance
- Knitted construction
- Nylon liner
- Water-based polyurethane and nitrile coating
- 3/4-Dipped finish
- Knitwrist cuff
Specifications
11-402 | |
Polyurethane | |
Nylon | |
Abrasions | |
11 | |
CE Mark, EN 388 : 4134 |
Polyurethane | |
Elasticated cuff | |
3/4 Dipped palm | |
11-402 | |
144 Pairs |