Learn More
Ansell HyFlex™ 11-130 Seamless Knit Polyamide Antistatic Gloves with Coated Palm
Gloves are knitted using a seamless, non-stop process with high-tenacity polyamid continuous thread. Antistatic properties protect users from electrostatic discharge.
Brand:  Ansell 11130070
Additional Details : Weight : 4.32000kg
 View more versions of this product
Product Code. 16019891
  • 44.30 EUR / 12 pairs
Estimated Shipment: 27-03-2023
to see stock.
Add to basket
Description
Description
  • Gloves are knitted using a seamless, non-stop process
  • Textile liner made of high-tenacity polyamid continuous thread that includes copper
  • Seamless, fine stitches enhance user comfort
  • Stretchable polyurethane coating on the palm for grip capabilities
  • Antistatic properties offer the user hand protection against electrostatic discharge
  • Low lint for cleanroom applications
Specifications
Specifications
EN 420:2003 + A1:2009
Polyurethane
Specific
Antistatic Gloves
3131A
Knit
11-130
Seamless Knit
Gray
Polyamide
7
Unisex
Coated
13