Learn More
Thermo Scientific™ Hydrogold Column Guard
Provides protection to the analytical column.
69.05 EUR - 111.22 EUR
Especificaciones
Diameter (Metric) Inner | 0.53 mm |
---|---|
Type | GC Guard Column |
Quantity | 1 |
Código de producto | Marca | Length (Metric) | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Length (Metric) | Precio | Cantidad y disponibilidad | |||||
15520312
|
Thermo Scientific™
26H50-0253 |
2 m |
69.05 EUR
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15560312
|
Thermo Scientific™
26H50-0553 |
5 m |
111.22 EUR
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Descripción
Protects against column contamination caused by non-volatile materials, extending the column lifetime.Especificaciones
0.53 mm | |
GC Guard Column | |
1 |