missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 25% w/w, Reagent Grade, Scharlab
Click to view available options
Quantity:
1 L
Description
- Reagent Grade
- Synonym: Hydrogen chloride solution
- Molecular Formula: HCl
- Molar Mass: 36.46g/mol
- CAS: 7647-01-0
- EIN ECS No.: 231-595-7
Specifications
CAS | 7647-01-0 |
Melting Point | ∼-70°C |
Boiling Point | 107°C |
Density | ∼1.12g/cm3 |
Residue after Ignition | 0.0005% max |
Vapor Pressure | 12hPa (at 20°C) |
Quantity | 1 L |
Molecular Formula | ClH |
Physical Form | Solution |
MDL Number | MFCD00011324 MFCD00792839 |
Show More |
Safety and Handling
Product Identifier
- Hydrochloric acid, solution 25% w/w, reagent grade, ISO
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Hydrogen chloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'