Learn More
HYDRANAL™-Humidity absorber, Honeywell Fluka™
Drying agent for air and gases for Karl Fischer titration (with indicator)
Brand: Honeywell Fluka 34788-6X500G
Additional Details : Weight : 0.50000kg
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
6 x 500 g | |
HYDRANAL™-Humidity absorber | |
NONH for all modes of transport |