missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRA-POINT™ Karl Fischer dry methanol (water content max. 0.01%), J.T. Baker™
Click to view available options
Quantity:
1 L
2.5 L
Description
- Volumetric reagents for water determination by Karl Fischer titration
- No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
- They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
- Formulated to reach each endpoint quickly, accurately and with reproducibility
- Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
- To be used with HYDRA-POINT™ Composite 2 product no. 8891 or HYDRA-POINT™ Composite 5 product no. 8890
Specifications
Flash Point | 11°C |
Quantity | 2.5 L |
Packaging | Glass bottle |
UN Number | 1230 |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'