missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human Transferrin protein
HumanKine™ recombinant protein
134.00 EUR - 2880.00 EUR
Specifications
Accession Number | P02787 |
---|---|
Formulation | 1 x PBS |
Gene ID (Entrez) | 7018 |
Molecular Weight (g/mol) | 70 kDa |
Source | HEK293 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17630077
|
Proteintech
HZ-1317-1000UG |
1000 μg |
2880.00 EUR
1000µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17640077
|
Proteintech
HZ-1317-100UG |
100 μg |
720.00 EUR
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17650077
|
Proteintech
HZ-1317-10UG |
10 μg |
134.00 EUR
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Transferrin is an 80kDA, monomeric glycoprotein that is responsible for binding ferric iron and transporting it throughout the body. It is primarily produced by hepatocytes and is a critical component of iron homeostasis as it delivers free iron to tissue sites where it is absorbed and utilized by the body. Once it reaches a site of absorption, iron-loaded transferrin binds to its corresponding transferrin receptor and taken up by the cell through endocytosis, after which the iron is disassociated, and free transferrin is released back into the bloodstream at a turnover rate of around 10 times per day. In addition to iron homeostasis, transferrin also plays an important role in the innate immune system. Downregulation of transferrin receptor expression has been linked to the reduced ability of intracellular pathogens to access a cell’s iron supply. Measurement of transferrin levels in the blood can also serve as a clinical marker for possible iron deficiency (PMID: 30422523, 22033148).Specifications
P02787 | |
7018 | |
HEK293 | |
<0.1 EU/μg | |
Recombinant | |
Lyophilized |
1 x PBS | |
70 kDa | |
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) | |
Human | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
>95% |