Learn More
Proteintech HumanKine™ recombinant human IL-1 alpha protein
Product Code. 17640087 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. 17640087
Brand: Proteintech HZ132010UG
HumanKine™ recombinant protein
Specifications
Accession Number P01583
Formulation 1 x PBS, See Certificate of Analysis for details
Gene ID (Entrez) 3552
Molecular Weight (g/mol) 22 kDa, monomer, glycosylated
Quantity 1000 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <1 EU/μg
Specific Reactivity Human|Mouse
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Activity 0.075-0.375 ng/mL EC50
Form Lyophilized
Purity or Quality Grade >95%