Learn More
Proteintech HumanKine™ recombinant human IGF-I protein
Product Code. 17690087 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. 17690087
Brand: Proteintech HZ1322100UG
HumanKine™ recombinant protein
The Insulin like growth factor-1, also known as Somatomedin-C is a growth factor which is structurally related to insulin and is an important regulator of growth and differentiation in various tissues and cell systems. Human IGF-1 is synthesized as two precursor isoforms with N- and alternate C-terminal propeptide. The two precursor isoforms are differentially expressed by various tissues. The proteolytic cleavage of the N- and C-terminal regions results in the mature IGF-1 protein which is identical between isoforms. IGF-1 binds to IGF-1 receptor and induces receptor autophosphorylation. This further phosphorylates Insulin receptor substrate -1 (IRS-1) and activates various downstream signaling pathways including the PI3-AKT, MAPK etc. (PMID: 17354613, 17113337, 29535161)
Specifications
Accession Number P05019
Formulation 1 x PBS pH 7.4
Gene ID (Entrez) 3479
Molecular Weight (g/mol) 10 kDa
Quantity 100 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Activity 2-14 ng/mL EC50
Form Lyophilized
Purity or Quality Grade >95%