Learn More
Abnova™ Human WARS2 Partial ORF (NP_056651.1, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_056651.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10352 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035235
|
Abnova™
H00010352-Q02.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025235
|
Abnova™
H00010352-Q02.10UG |
10 ug |
335.00 EUR
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described. [provided by RefSeq]
Sequence: MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQSpecifications
NP_056651.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TrpRS | |
WARS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10352 | |
WARS2 (Human) Recombinant Protein (Q02) | |
MALHSMRKARERWSFIRALHKGSAAAPALQKDSKKRVFSGIQPTGILHLGNYLGAIESWVRLQDEYDSVLYSIVDLHSITVPQDPAVLRQ | |
RUO | |
WARS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |