Learn More
Abnova™ Human TLN1 Partial ORF (AAH42923.1, 86 a.a. - 1430 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007094-Q06.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a cytoskeletal protein that is concentrated in areas of cell-substratum and cell-cell contacts. The encoded protein plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. It codistributes with integrins in the cell surface membrane in order to assist in the attachment of adherent cells to extracellular matrices and of lymphocytes to other cells. The N-terminus of this protein contains elements for localization to cell-extracellular matrix junctions. The C-terminus contains binding sites for proteins such as beta-1-integrin, actin, and vinculin. [provided by RefSeq]
Sequence: RPLKIRMLDGTVKTIMVDDSKTVTDMLMTICARIGITNHDEYSLVRELMEEKKEEITGTLRKDKTLLRDEKKMEKLKQKLHTDDELNWLDHGRTLREQGVEEHETLLLRRKFFYSDQNVDSRDPVQLNLLYVQARDDILNGSHPVSFDKACEFAGFQCQIQFGPHNEQKHKAGFLDLKDFLPKEYVKQKGERKIFQAHKNCGQMSEIEAKVRYVKLARSLKTYGVSFFLVKEKMKGKNKLVPRLLGITKECVMRVDEKTKEVIQEWNLTNIKRWAASPKSFTLDFGDYQDGYYSVQTTEGEQIAQLIAGYIDIILKKKKSKDHFGLEGDEESTMLEDSVSPKKSTVLQQQYNRVGKVEHGSVALPAIMRSGASGPENFQVGSMPPAQQQITSGQMHRGHMPPLTSAQQALTGTINSSMQAVQAAQATLDDFDTLPPLGQDAASKAWRKNKMDESKHEIHSQVDAITAGTASVVNLTAGDPAETDYTAVGCAVTTISSNLTEMSRGVKLLAALLEDEGGSGRPLLQAAKGLAGAVSELLRSAQPASAEPRQNLLQAAGNVGQASGELLQQIGESDTDPHFQDALMQLAKAVASAAAALVLKAKSVAQRTEDSGLQTQVIAAATQCALSTSQLVACTKVVAPTISSPVCQEQLVEAGRLVAKAVEGCVSASQAATEDGQLLRGVGAAATAVTQALNELLQHVKAHATGAGPAGRYDQATDTILTVTENIFSSMGDAGEMVRQARILAQATSDLVNAIKADAEGESDLENSRKLLSAAKILADATAKMVEAAKGAAAHPDSEEQQQRLREAAEGLRMATNAAAQNAIKKKLVQRLEHAAKQAAASATQTIAAAQHAASTPKASAGPQPLLVQSCKAVAEQIPLLVQGVRGSQAQPDSPSAQLALIAASQSFLQPGGKMVAAAKASVPTIQDQASAMQLSQCAKNLGTALAELRTAAQKAQEACGPLEMDSALSVVQNLEKDLQEVKAAARDGKLKPLPGETMEKCTQDLGNSTKAVSSAIAQLLGEVAQGNENYAGIAARDVAGGLRSLAQAARGVAALTSDPAVQAIVLDTASDVLDKASSLIEEAKKAAGHPGDPESQQRLAQVAKAVTQALNRCVSCLPGQRDVDNALRAVGDASKRLLSDSLPPSTGTFQEAQSRLNEAAAGLNQAATELVQASRGTPQDLARASGRFGQDFSTFLEAGVEMAGQAPSQEDRAQVVSNLKGISMSSSKLLLAAKALSTDPAAPNLKSQLAAAARAVTDSINQLITMCTQQAPGQKECDNALRELETVRELLENPVQPINDMSYFGCLDSVMENSKVLGEAMTGISQNAKNGNLPEFGDAISTASSpecifications
AAH42923.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
173.58kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
RPLKIRMLDGTVKTIMVDDSKTVTDMLMTICARIGITNHDEYSLVRELMEEKKEEITGTLRKDKTLLRDEKKMEKLKQKLHTDDELNWLDHGRTLREQGVEEHETLLLRRKFFYSDQNVDSRDPVQLNLLYVQARDDILNGSHPVSFDKACEFAGFQCQIQFGPHNEQKHKAGFLDLKDFLPKEYVKQKGERKIFQAHKNCGQMSEIEAKVRYVKLARSLKTYGVSFFLVKEKMKGKNKLVPRLLGITKECVMRVDEKTKEVIQEWNLTNIKRWAASPKSFTLDFGDYQDGYYSVQTTEGEQIAQLIAGYIDIILKKKKSKDHFGLEGDEESTMLEDSVSPKKSTVLQQQYNRVGKVEHGSVALPAIMRSGASGPENFQVGSMPPAQQQITSGQMHRGHMPPLTSAQQALTGTINSSMQAVQAAQATLDDFDTLPPLGQDAASKAWRKNKMDESKHEIHSQVDAITAGTASVVNLTAGDPAETDYTAVGCAVTTISSNLTEMSRGVKLLAALLEDEGGSGRPLLQAAKGLAGAVSELLRSAQPASAEPRQNLLQAAGNVGQASGELLQQIGESDTDPHFQDALMQLAKAVASAAAALVLKAKSVAQRTEDSGLQTQVIAAATQCALSTSQLVACTKVVAPTISSPVCQEQLVEAGRLVAKAVEGCVSASQAATEDGQLLRGVGAAATAVTQALNELLQHVKAHATGAGPAGRYDQATDTILTVTENIFSSMGDAGEMVRQARILAQATSDLVNAIKADAEGESDLENSRKLLSAAKILADATAKMVEAAKGAAAHPDSEEQQQRLREAAEGLRMATNAAAQNAIKKKLVQRLEHAAKQAAASATQTIAAAQHAASTPKASAGPQPLLVQSCKAVAEQIPLLVQGVRGSQAQPDSPSAQLALIAASQSFLQPGGKMVAAAKASVPTIQDQASAMQLSQCAKNLGTALAELRTAAQKAQEACGPLEMDSALSVVQNLEKDLQEVKAAARDGKLKPLPGETME | |
RUO | |
TLN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7094 | |
TLN1 (Human) Recombinant Protein (Q06) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ILWEQ/KIAA1027/TLN | |
TLN1 | |
Recombinant | |
wheat germ expression system |