missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SSH1 Partial ORF (NP_061857.2, 752 a.a. - 849 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 EUR - 508.00 EUR

Specifications

Accession Number NP_061857.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54434
Molecular Weight (g/mol) 36.52kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16055535
View Documents Promotion Details
Abnova™
H00054434-Q02.25UG
25 ug
508.00 EUR
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16045535
View Documents Promotion Details
Abnova™
H00054434-Q02.10UG
10 ug
335.00 EUR
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM]

Sequence: PKSLLLKNSHCDKNPPSTEVVIKEESSPKKDMKPAKDLRLLFSNESEKPTTNSYLMQHQESIIQLQKAGLVRKHTKELERLKSVPADPAPPSRDGPAS
Specifications

Specifications

NP_061857.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38102/KIAA1298/SSH-1
SSH1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
54434
SSH1 (Human) Recombinant Protein (Q02)
PKSLLLKNSHCDKNPPSTEVVIKEESSPKKDMKPAKDLRLLFSNESEKPTTNSYLMQHQESIIQLQKAGLVRKHTKELERLKSVPADPAPPSRDGPAS
RUO
SSH1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications