missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RP1-127L4.6 Full-length ORF (AAI39845.2, 1 a.a. - 256 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | AAI39845.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 150297 |
Molecular Weight (g/mol) | 54.56kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16168603
|
Abnova™
H00150297-P01.10UG |
10 ug |
335.00 EUR
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178603
|
Abnova™
H00150297-P01.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MGSKLTCCLGPSGGLNCDCCRPDVGPCHECEIPETVAATAPASTTAKPAKLDLKAKKAQLMQYLSLPKTPKMLKMSKGLDARSKRWLKIIWRRHGIWPLENIGPTEDVQASAHGGVEENMTSDIEIPEAKHDHRPTEDVQVSAHGGVEENITSDIEISEAKHDHHLVEDLSESLSVCLEDFMTSDLSESLSVSLEDFMTSGLSESLSVSLEDLMTPEMAKERYEDYLCWVKMARSRLNEPISSQVLGLLRLPTFLYSpecifications
AAI39845.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.56kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGSKLTCCLGPSGGLNCDCCRPDVGPCHECEIPETVAATAPASTTAKPAKLDLKAKKAQLMQYLSLPKTPKMLKMSKGLDARSKRWLKIIWRRHGIWPLENIGPTEDVQASAHGGVEENMTSDIEIPEAKHDHRPTEDVQVSAHGGVEENITSDIEISEAKHDHHLVEDLSESLSVCLEDFMTSDLSESLSVSLEDFMTSGLSESLSVSLEDLMTPEMAKERYEDYLCWVKMARSRLNEPISSQVLGLLRLPTFLY | |
RUO | |
RP1-127L4.6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
150297 | |
RP1-127L4.6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LOC150297/dJ90G24.6 | |
RP1-127L4.6 | |
Recombinant | |
wheat germ expression system |