Learn More
Invitrogen™ Human ROR1 ELISA Kit
Human ROR1 quantitates human ROR1 in serum, plasma, supernatant.
Brand: Invitrogen™ EH395RB
Additional Details : Weight : 0.33820kg
Description
Human ROR1 quantitates human ROR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ROR1.
ROR1 is a receptor protein tyrosine kinase whose cellular role has not been determined. It is a type I membrane protein and belongs to the ROR subfamily of cell surface receptors. Studies of a similar protein in mouse suggest that this protein may interact with another receptor protein tyrosine kinase and may be involved in skeletal and cardiac development.Specifications
2810404D04Rik; dJ537F10.1; Inactive tyrosine-protein kinase transmembrane receptor ROR1; MGC99659; mRor1; neurotrophic tyrosine kinase, receptor related 1; neurotrophic tyrosine kinase, receptor-related 1; Ntrkr1; receptor tyrosine kinase like orphan receptor 1; receptor tyrosine kinase-like orphan receptor 1; RGD1559469; Ror1; T-ROR1; tyrosine-protein kinase transmembrane receptor ROR1 | |
0.82 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
ROR1 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.819 to 200 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q01973 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
ROR1 | |
4 hr 45 min |