Learn More
Novus Biologicals™ Human Presenilin-1 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand:  Novus Biologicals™ NBP3-06975
Product Code. 18331763
  • 814.00 EUR / 1 set
Specifications
0.31 to 20 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human Presenilin-1 in samples. No significant cross-reactivity or interference between Human Presenilin-1 and analogues was observed.
5663
1 Kit
Storage of components varies. See protocol for specific instructions.
Human
0.19 ng/mL
Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
PSEN1
Alzheimers Research, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers, Stem Cell Signaling Pathway, Tumor Suppressors, Wnt Signaling Pathway
AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182