Learn More
Novus Biologicals™ Human Presenilin-1 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand: Novus Biologicals™ NBP3-06975
Additional Details : Weight : 0.00970kg
Specifications
0.31 to 20 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human Presenilin-1 in samples. No significant cross-reactivity or interference between Human Presenilin-1 and analogues was observed. | |
5663 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |
0.19 ng/mL | |
Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
PSEN1 | |
Alzheimers Research, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers, Stem Cell Signaling Pathway, Tumor Suppressors, Wnt Signaling Pathway | |
AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182 |