missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEUROD6 Partial ORF (NP_073565.2, 191 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_073565.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 63974 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035625
|
Abnova™
H00063974-Q03.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025625
|
Abnova™
H00063974-Q03.10UG |
10 ug |
335.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM]
Sequence: GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMHSpecifications
NP_073565.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Atoh2/MATH2/Math-2/NEX1M/bHLHa2 | |
NEUROD6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
63974 | |
NEUROD6 (Human) Recombinant Protein (Q03) | |
GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMH | |
RUO | |
NEUROD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |