missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYO3B Partial ORF (NP_620482.1, 280 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 EUR - 508.00 EUR

Specifications

Accession Number NP_620482.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 140469
Molecular Weight (g/mol) 35.64kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16025785
View Documents Promotion Details
Abnova™
H00140469-Q01.10UG
10 ug
335.00 EUR
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16035785
View Documents Promotion Details
Abnova™
H00140469-Q01.25UG
25 ug
508.00 EUR
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: RRPSVTHLLDHPFIKGVHGKVLFLQKQLAKVLQDQKHQNPVAKTRHERMHTRRPYHVEDAEKYCLEDDLVNLEVLDEDTIIHQLQKRYAD
Specifications

Specifications

NP_620482.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MYO3B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
140469
MYO3B (Human) Recombinant Protein (Q01)
RRPSVTHLLDHPFIKGVHGKVLFLQKQLAKVLQDQKHQNPVAKTRHERMHTRRPYHVEDAEKYCLEDDLVNLEVLDEDTIIHQLQKRYAD
RUO
MYO3B
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications