missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human Mycoplasma pneumoniae Antibody (IgG) ELISA Kit (Colorimetric)
Shop All Bio Techne ProductsDescription
The Human Mycoplasma pneumoniae Antibody (IgG) ELISA Kit (Colorimetric) measures human Mycoplasma pneumoniae Antibody (IgG) in serum.
Specifications
Specifications
| Assay Range | Qualitative |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Research Discipline | Cell Biology |
| Sample Volume | 50 to 100 μL |
| Storage Requirements | Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?