Learn More
Abnova™ Human MTA1 Partial ORF (NP_003122, 542 a.a. - 632 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009112-Q03.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling. [provided by RefSeq]
Sequence: ETHPRPPKPDPVKSVSSVLSSLTPAKVAPVINNGSPTILGKRSYEQHNGVDGNMKKRLLMPSRGLANHGQTRHMGPSRNLLLNGKSYPTKVSpecifications
NP_003122 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
ETHPRPPKPDPVKSVSSVLSSLTPAKVAPVINNGSPTILGKRSYEQHNGVDGNMKKRLLMPSRGLANHGQTRHMGPSRNLLLNGKSYPTKV | |
RUO | |
MTA1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9112 | |
MTA1 (Human) Recombinant Protein (Q03) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MTA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |