missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MARCH9 Partial ORF (NP_612405.2, 240 a.a. - 345 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_612405.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 92979 |
Molecular Weight (g/mol) | 37.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035735
|
Abnova™
H00092979-Q01.10UG |
10 ug |
335.00 EUR
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045735
|
Abnova™
H00092979-Q01.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: HEGSSVYRIFKRWQAVNQQWKVLNYDKTKDIGGDAGGGTAGKSGPRNSRTGPTSGATSRPPAAQRMRTLLPQRCGYTILHLLGQLRPPDARSSSHSGREVVMRVTTSpecifications
NP_612405.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ36578/MARCH-IX/RNF179 | |
MARCH9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
92979 | |
MARCH9 (Human) Recombinant Protein (Q01) | |
HEGSSVYRIFKRWQAVNQQWKVLNYDKTKDIGGDAGGGTAGKSGPRNSRTGPTSGATSRPPAAQRMRTLLPQRCGYTILHLLGQLRPPDARSSSHSGREVVMRVTT | |
RUO | |
MARCH9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |