Learn More
Novus Biologicals™ Human IL-10 R beta ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand:  Novus Biologicals™ NBP3-06792
Product Code. 18331772
  • 814.00 EUR / 1 set
Specifications
31.25 to 2000 pg/mL
ELISA Kit (Colorimetric)
This kit recognizes Human IL-10 R beta in samples. No significant cross-reactivity or interference between Human IL-10 R beta and analogues was observed.
3588
1 Kit
Storage of components varies. See protocol for specific instructions.
Human
18.75 pg/mL
Biological Fluids, Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
IL10RB
Cytokine Research, Immunology, Innate Immunity
CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4