missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GZMH Partial ORF (NP_219491, 161 a.a. - 246 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_219491 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2999 |
Molecular Weight (g/mol) | 35.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034105
|
Abnova™
H00002999-Q02.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024105
|
Abnova™
H00002999-Q02.10UG |
10 ug |
335.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence: EVLLTVQKDCQCERLFHGNYSRATEICVGDPKKTQTGFKGDSGGPLVCKDVAQGILSYGNKKGTPPGVYIKVSHFLPWIKRTMKRLSpecifications
NP_219491 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCP-X/CGL-2/CSP-C/CTLA1/CTSGL2 | |
GZMH | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2999 | |
GZMH (Human) Recombinant Protein (Q02) | |
EVLLTVQKDCQCERLFHGNYSRATEICVGDPKKTQTGFKGDSGGPLVCKDVAQGILSYGNKKGTPPGVYIKVSHFLPWIKRTMKRL | |
RUO | |
GZMH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |