Learn More
Invitrogen™ Human Glucocorticoid Receptor beta (aa 728-742) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP222
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human GR beta (residues 728-742) Synthetic Peptide, PEP-222, from Invitrogen.
PEP-222 is a 16 amino acid synthetic peptide corresponding to amino acid residues 728-742 from human GR beta. The sequence of this peptide is (amino to carboxy terminus): N (728) V M W L K P E S T S H T L I (742) CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA3-514. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
2908 | |
purified | |
Human Glucocorticoid Receptor beta (aa 728-742) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P04150 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
NR3C1 | |
No preservative | |
50 μg | |
N(728) V M W L K P E S T S H T L I(742) C |