missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GDF7 Partial ORF (NP_878248.2, 204 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_878248.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 151449 |
Molecular Weight (g/mol) | 36.41kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015805
|
Abnova™
H00151449-Q02.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005805
|
Abnova™
H00151449-Q02.10UG |
10 ug |
335.00 EUR
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord. [provided by RefSeq]
Sequence: VGQRWEAFDVADAMRRHRREPRPPRAFCLLLRAVAGPVPSPLALRRLGFGWPGGGGSAAEERAVLVVSSRTQRKESLFREIRAQARALGAALASEPLPSpecifications
NP_878248.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMP12 | |
GDF7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
151449 | |
GDF7 (Human) Recombinant Protein (Q02) | |
VGQRWEAFDVADAMRRHRREPRPPRAFCLLLRAVAGPVPSPLALRRLGFGWPGGGGSAAEERAVLVVSSRTQRKESLFREIRAQARALGAALASEPLP | |
RUO | |
GDF7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |