missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®

Description
Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
Specifications
Specifications
| Accession Number | P21781 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2252 |
| Molecular Weight (g/mol) | 18.9 kDa |
| Name | Human FGF-7 (KGF), Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 2 μg |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?