missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DEAF1 Partial ORF (NP_066288.2, 133 a.a. - 222 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010522-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: TALQIGDSLNTEKATLIVVHTDGSIVETTGLKGPAAPLTPGPQSPPTPLAPGQEKGGTKYNWDPSVYDSELPVRCRNISGTLYKNRLGSGSpecifications
NP_066288.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TALQIGDSLNTEKATLIVVHTDGSIVETTGLKGPAAPLTPGPQSPPTPLAPGQEKGGTKYNWDPSVYDSELPVRCRNISGTLYKNRLGSG | |
RUO | |
DEAF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10522 | |
DEAF1 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NUDR/SPN/ZMYND5 | |
DEAF1 | |
Recombinant | |
wheat germ expression system |