missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cytokeratin 19/KRT19 ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EH146RBX10

Additional Details : Weight : 2.50000kg

Product Code. 17717573

  • 5100.00 EUR / Pack of 10
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.
Specifications

Specifications

P08727
0.061 ng/mL
ELISA
Colorimetric Microplate Reader
KRT19
<10%
HRP
RUO
2°C to 8°C
1 hr 20 min
0.061 to 15 ng/mL
Biotin
Plasma, Serum, Supernatant
3880
<12%
Pre-coated 96 well plate|Standard|Assay Diluent concentrate|Biotinylated Detection Antibody|SAV-HRP|Wash Buffer|Chromogen|Stop Solution|Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL|Serum, 50 μL|Supernatant, 50 μL
Cytokeratin 19
4 hr 45 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers