Learn More
Novus Biologicals™ Human Cytokeratin 13 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75282
Product Code. 18331351
  • 836.00 EUR / 1 set
Description
The Human Cytokeratin 13 ELISA Kit (Colorimetric) measures human Cytokeratin 13 in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
This kit recognizes Human CK-13/KRT13 in samples. No significant cross-reactivity or interference between Human CK-13/KRT13 and analogues was observed.
Biological Fluids, Plasma, Serum
3860
Human
Storage of components varies. See protocol for specific instructions.
9.38 pg/mL
ELISA Kit (Colorimetric)
ELISA
KRT13
1 Kit
Cytokeratin 13