Learn More
Novus Biologicals™ Human Cytokeratin 13 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75282
Additional Details : Weight : 0.00970kg
Description
The Human Cytokeratin 13 ELISA Kit (Colorimetric) measures human Cytokeratin 13 in serum, plasma and other biological fluids.Specifications
15.63 - 1000 pg/mL | |
This kit recognizes Human CK-13/KRT13 in samples. No significant cross-reactivity or interference between Human CK-13/KRT13 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
3860 | |
Human | |
Storage of components varies. See protocol for specific instructions. |
9.38 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
KRT13 | |
1 Kit | |
Cytokeratin 13 |