missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human CXCL7/NAP-2 ELISA Kit (Colorimetric)

Product Code. 18214943 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214943 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214943 Supplier Novus Biologicals™ Supplier No. NBP2599651KIT
  • ONLINE EXCLUSIVE PRICE
    764.00 EUR
    722.40 EUR / 1 sets
    Save 41.60 EUR
    5% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Assay has high sensitivity and specificity for detection of human NAP-2

The Human CXCL7/NAP-2 ELISA Kit (Colorimetric) measures human CXCL7/NAP-2 in serum, plasma, tissue homogenates.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78pg/ml to 5000pg/ml
Assay Sensitivity 19.5pg/mL.
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity For detection of human NAP-2
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5473
Gene Symbol PPBP
Kit Contents Assay plate (12 x 8 coated Microwells), Biotin-antibody (100 x concentrate), Biotin-antibody Diluent, Adhesive Strip (For 96 wells), HRP-avidin Diluent, HRP-avidin (100 x concentrate), TMB Substrate, Wash Buffer (25 x concentrate), Standard (Freeze dried), Sample Diluent, Stop Solution, Instruction manual
Quantity 1 Kit
Research Discipline Immunology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym Beta-TG, connective tissue-activating peptide III, CTAP3CTAP-III, CTAPIII, CXC chemokine ligand 7, C-X-C motif chemokine 7, CXCL7b-TG1, LA-PF4, LDGFTC1, Leukocyte-derived growth factor, low-affinity platelet factor IV, Macrophage-derived growth factor, MDGFTC2, NAP-2, NAP-2-L1, neutrophil-activating peptide 2, neutrophil-activating peptide-2, PBPTHBGB, platelet basic protein, pro-platelet basic protein (chemokine (C-X-C motif) ligand 7), SCAR10, SCYB7beta-thromboglobulin, small inducible cytokine B7, small inducible cytokine subfamily B, member 7, Small-inducible cytokine B7, TGB, TGB1B-TG1, THBGB1, thrombocidin 1, thrombocidin 2, thromboglobulin, beta-1
Target CXCL7/NAP-2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.