Learn More
R&D Systems™ Human CTGF/CCN2 Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 9190-CC-050
Additional Details : Weight : 0.00970kg
Description
Measured by its ability to mediate Balb/3T3 mouse embryonic fibroblast cell adhesion. Ball, D.K. et al. (2003) Reproduction 125:271. The ED50 for this effect is1.5-7.5 μg/mL.Order Info
The product is shipped at ambient temperature.
Specifications
Q5M8T4 | |
1490 | |
CCN2, CCN2IGFBP-8, connective tissue growth factor, CTGRP, Fisp12, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP-8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
Mouse myeloma cell line,NS0-derived human CTGF/CCN2 protein Gln27-Ala349 | |
RUO | |
1.5 to 7.5μg/mL (ED 50) | |
Unconjugated |
Lyophilized from a 0.2μm filtered solution in PBS. | |
35 kDa, reducing conditions | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
<0.10 EU per 1μg of protein by LAL method | |
Recombinant protein | |
Yes |
For Research Use Only