Learn More
Novus Biologicals™ Human Chymotrypsin C/CTRC ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand: Novus Biologicals™ NBP3-06736
This item is not returnable.
View return policy
Specifications
3.13 to 200 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human Elastase in samples. No significant cross-reactivity or interference between Human Elastase and analogues was observed. | |
11330 | |
1 Kit | |
Caldecrin, chymotrypsin C (caldecrin), chymotrypsin-C, CLCRserum calcium decreasing factor, EC 3.4.21, EC 3.4.21.2, ELA4, elastase 4, elastase IV |
1.88 ng/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
CTRC | |
Storage of components varies. See protocol for specific instructions. | |
Human |