missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF2 Partial ORF (NP_004714, 590 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_004714 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9181 |
Molecular Weight (g/mol) | 36.85kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045135
|
Abnova™
H00009181-Q02.10UG |
10 ug |
335.00 EUR
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055135
|
Abnova™
H00009181-Q02.25UG |
25 ug |
508.00 EUR
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate rho-dependent signals. Alternatively spliced transcript variants encoding different isoforms have been identified.[provided by RefSeq]
Sequence: MTHFQAEEDGGSGMALPTLPRGLFRSESLESPRGERLLQDAIREVEGLKDLLVGPGVELLLTPREPALPLEPDSGGNTSPGVTANGEARTFNGSIELCRADSpecifications
NP_004714 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp547L106/DKFZp547P1516/GEF/GEF-H1/GEFH1/KIAA0651/LFP40/P40 | |
ARHGEF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9181 | |
ARHGEF2 (Human) Recombinant Protein (Q02) | |
MTHFQAEEDGGSGMALPTLPRGLFRSESLESPRGERLLQDAIREVEGLKDLLVGPGVELLLTPREPALPLEPDSGGNTSPGVTANGEARTFNGSIELCRAD | |
RUO | |
ARHGEF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |