Learn More
Abnova™ Human AKAP7 Partial ORF (NP_057461.1, 2 a.a. - 115 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009465-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the A-kinase anchoring protein (AKAP) family, a group of functionally related proteins that bind to a regulatory subunit (RII) of cAMP-dependent protein kinase A (PKA) and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined. [provided by RefSeq]
Sequence: SEEFEANTMDSLVDMPFATVDIQDDCGITDEPQINLKRSQENEWVKSDQVKKRKKKRKDYQPNYFLSIPITNKEIIKGIKILQNAIIQQDERLAKAMVSDGSFHITLLVMQLLNSpecifications
NP_057461.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.28kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEEFEANTMDSLVDMPFATVDIQDDCGITDEPQINLKRSQENEWVKSDQVKKRKKKRKDYQPNYFLSIPITNKEIIKGIKILQNAIIQQDERLAKAMVSDGSFHITLLVMQLLN | |
RUO | |
AKAP7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9465 | |
AKAP7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AKAP18 | |
AKAP7 | |
Recombinant | |
wheat germ expression system |