missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Hubtron™ Pascal Fluoroelastomer Tubing

Ideal for extremely demanding corrosive chemical transfer, electronic applications, ink and general printing applications, automotive applications, general mechanical applications

Brand:  Cole-Parmer™ 95227-02

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11745308

Quantity Price
1 658.00 EUR / Each
5 579.11 EUR / Each
Estimated Shipment
11-05-2021
Add to basket


Description

Description

  • Superior pressure ratings.
  • Extremely strong chemical resistance.
  • Extremely low permeability.
  • Excellent resistance to solvents, acids, bases, alcohols and esters.
  • Autoclavable.
Specifications

Specifications

Resin: 3A;ADCF;FDA;Non-DEHP;Non-Phthalate;European Pharmacopoeia;NSF-51;USP;NSF-61
248°F
Autoclavable
PTFE
25 ft.
1.6 mm
-10°C
Transparent
120°C
3.2 mm
6.4 mm
7.6 m
14°F
Videos
SDS
Documents