missing translation for 'onlineSavingsMsg'
Learn More

Honeywell KCL™ Honeywell KCL™ Camper™ Comfort Gloves

Catalog Number 15487170
Click to view available options
Size:
7
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from polyurethane. Honeywell KCL™ Honeywell KCL™ Camper™ Comfort Gloves offers high mechanical resistance and maximum touch sensitivity. The polyurethane gives the glove a very good grip, even on wet and oily objects.

  • Washable marked according to size with different colors on the wristband
    Excellent fit very good sensitivity
    Very breathable
    Good resistance to mechanical damage
    Excellent grip when handling greasy and oily parts
TRUSTED_SUSTAINABILITY
Certifications/Compliance 0075/1445/162/06/12/0481
Cuff Material Polyurethane
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Polyurethane
Powdered Powder Free
Size 7
Sterility Non-sterile
Thickness (Metric) 1.0 cm
Glove Cuff Color White
Length (Metric) 220 to 260 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'