Learn More
Hoefer™ Low Fluorescence Glass Plates
For use with SE660, SE600X Chroma, SE600, SE640, SE410 and SE400
Brand:  Hoefer™ SE6602LF
Additional Details : Weight : 0.80000kg
Product Code. 11955378
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • 18 x 24 cm
  • 2 pcs
Specifications
Specifications
SE660, SE600X Chroma, SE600, SE640, SE410 and SE400 units
Low Fluorescence Glass
24 cm
18 cm