Learn More
Hexaammineruthenium(II) chloride, 99.9%, Thermo Scientific Chemicals
Catalog Number p-7027265
Click to view available options
Quantity:
0.25 g
1 g
15305-72-3
ClH12N6Ru
232.66
MFCD00011477
MHYZHXDLILNYTQ-UHFFFAOYSA-M
triton rw 30, azane;ruthenium 2+ ;dichloride, ruthenium iii chloride hexaammoniate, hexaammineruthenium ii chloride
15305-72-3
ClH12N6Ru
232.66
MFCD00011477
MHYZHXDLILNYTQ-UHFFFAOYSA-M
triton rw 30, azane;ruthenium 2+ ;dichloride, ruthenium iii chloride hexaammoniate, hexaammineruthenium ii chloride
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Odor Odorless
Quantity 0.25 g
Sensitivity Air sensitive
Formula Weight 274.16
Percent Purity 99.9%
Chemical Name or Material Hexaammineruthenium(II) chloride