Learn More
Thermo Scientific™ Herasafe™ KS Options and Accessories
Worksurfaces, exhaust transitions and baskets
Brand:  Thermo Scientific™ 50078795
650.09 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 10398179
  • 889.00 EUR / Each
Estimated Shipment: 01-08-2024
Description
Worksurfaces, exhaust transitions and baskets
Specifications
Specifications
Thimble Duct Exhaust Transition for Herasafe KS 18 (6 ft) Cabinet
Thimble Duct Exhaust Transition Accessory
Biosafety Cabinet