Learn More
Thermo Scientific™ Heracell™ Split Shelf System
Material: Copper; For Use With: Heracell™ VIOS CO2 Incubators
Brand: Thermo Scientific™ 51901122
Additional Details : Weight : 0.10000kg
Description
Half shelf system, Split Shelf; 6 half-width in place of 3 full -width, Material: copper; For use with Heracell VIOS 250i CO2 Incubator
- For use with 6 gas tight inner door configuration
- Half shelf system
- 6 half-width in place of 3 full-width
Specifications
6 split copper shelves | |
LED | |
Accessory, Shelf System |
Heracell™ VIOS CO2 Incubators | |
Copper |