missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin A1c Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen R01081D
Additional Details : Weight : 0.99000kg
Description
Purity: IgG is > 99% total protein by SDS-PAGE.
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.Specifications
Hemoglobin A1c | |
Polyclonal | |
Unconjugated | |
HBA1 | |
alpha one globin; alpha-2 globin chain; Alpha-globin; delta globin; HBA1; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3039 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunodiffusion, Immunoelectrophoresis, Western Blot | |
1.0 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
P69905 | |
HBA1 | |
Hemoglobin A1c | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |