Learn More
Keystone Cleanroom Products Heat-Seal Tyvek Autoclave Bag
GREENER_CHOICE
Product Code. 17708167 Shop All Keystone Cleanroom Products Products
Click to view available options
Unit Size:
Pack of 100
Pack of 50
Dimensions (L x W):
26 x 16 in.
30 x 27 in.
Product Code. 17708167
Brand: Keystone Cleanroom Products BGT2A4236MITT2WOCPE
Only null left
Add to basket
Add to basket
Keystone Cleanroom Products Heat-Seal Tyvek Autoclave Bag is a portable isolator in an easy to use, economical bag form. Ideal for difficult-to-sterilize equipment that typically requires aseptic connections after sterilization.
  • Tyvek™/PE Film construction for excellent strength and exceptional microbial barrier
  • Designed for steam sterilization
  • Heat Seal Closure
  • Integrated Mittens
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Make aseptic connections inside the Isolator Bag
  • Eliminates risks of contamination
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Includes Integrated mittens, see-through window, and steam indicator
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Sterilization Method Steam
Length (English) 36 in.
Length (Metric) 91.4 cm
Width (English) 42 in.
Width (Metric) 106.7 cm
Product Type Autoclave Bag