Learn More
Kappes Systeme™ HDPE Storage Bin
Brand: Kappes Systeme™ 6005.00.0552
Additional Details : Weight : 0.10000kg
Specifications
Storage Bin | |
Yellow | |
Feed and Roller Conveyor | |
7 | |
160 x 105 x 75 mm | |
105 mm |
HDPE | |
Rectangular | |
Food Storage | |
1/Pk. | |
160 mm | |
75 mm |